Assay Details
| Gene Symbol: cox4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214701.1 | NP_999866.1 | 2 - 3 | 112 | 71 | |
| GenBank mRNA | AY391417.1 | - | 2 - 3 | 112 | 71 | |
| BC095163.1 | - | 2 - 3 | 71 | |||
| BC164879.1 | - | 1 - 2 | 71 | |||
More Information
Set Membership: |
|

cox4i1
Genomic Map