Assay Details
| Gene Symbol: hspa4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214716.1 | NP_999881.1 | 12 - 13 | 1745 | 55 | |
| GenBank mRNA | BC051152.1 | - | 12 - 13 | 1745 | 55 | |
| BC165369.1 | - | 12 - 13 | 1595 | 55 | ||
More Information
Set Membership: |
|

hspa4a
Genomic Map