Assay Details
| Gene Symbol: irf2bp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214752.1 | NP_999917.1 | 1 - 2 | 940 | 77 | |
| GenBank mRNA | BC054562.1 | - | 1 - 2 | 940 | 77 | |
More Information
Set Membership: |
|

irf2bp2b
Genomic Map