Assay Details
| Gene Symbol: acp5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214773.1 | NP_999938.1 | 2 - 3 | 127 | 140 | |
| XM_005165725.4 | XP_005165782.1 | 2 - 3 | 135 | 140 | ||
| XM_005165726.4 | XP_005165783.1 | 2 - 3 | 212 | 140 | ||
| GenBank mRNA | BC055256.1 | - | 2 - 3 | 127 | 140 | |
| HM239646.1 | - | 1 - 2 | 90 | 140 | ||
More Information
Set Membership: |
|

acp5a
Genomic Map