Assay Details
| Gene Symbol: psmd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002112.2 | NP_001002112.1 | 6 - 7 | 669 | 113 | |
| GenBank mRNA | BC071482.1 | - | 6 - 7 | 670 | 113 | |
More Information
Set Membership: |
|

psmd4a
Genomic Map