Assay Details
| Gene Symbol: psmd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002112.2 | NP_001002112.1 | 8 - 9 | 917 | 91 | |
| GenBank mRNA | BC071482.1 | - | 8 - 9 | 918 | 91 | |
More Information
Set Membership: |
|

psmd4a
Genomic Map