Assay Details
| Gene Symbol: rpp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002148.1 | NP_001002148.1 | 6 - 7 | 825 | 81 | |
| GenBank mRNA | BC071381.1 | - | 6 - 7 | 825 | 81 | |
More Information
Set Membership: |
|

rpp40
Genomic Map