Assay Details
| Gene Symbol: calua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002151.2 | NP_001002151.2 | 4 - 5 | 751 | 152 | |
| NM_001317392.1 | NP_001304321.1 | 4 - 5 | 741 | 152 | ||
| GenBank mRNA | BC071361.1 | - | 4 - 5 | 694 | 152 | |
| EH487264.1 | - | 4 - 5 | 663 | 152 | ||
| EH505062.1 | - | 4 - 5 | 725 | 152 | ||
| EH588521.1 | - | 4 - 5 | 721 | 152 | ||
| KM655576.1 | - | 3 - 4 | 589 | 152 | ||
More Information
Set Membership: |
|

calua
Genomic Map