Assay Details
| Gene Symbol: calua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002151.2 | NP_001002151.2 | 5 - 6 | 810 | 103 | |
| NM_001317392.1 | NP_001304321.1 | 5 - 6 | 800 | 103 | ||
| GenBank mRNA | BC071361.1 | - | 5 - 6 | 753 | 103 | |
| EH487264.1 | - | 5 - 6 | 722 | 103 | ||
| EH505062.1 | - | 5 - 6 | 784 | 103 | ||
| EH588521.1 | - | 5 - 6 | 780 | 103 | ||
| KM655576.1 | - | 4 - 5 | 648 | 103 | ||
More Information
Set Membership: |
|

calua
Genomic Map