Assay Details
| Gene Symbol: calua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002151.2 | NP_001002151.2 | 6 - 7 | 1016 | 89 | |
| NM_001317392.1 | NP_001304321.1 | 6 - 7 | 1006 | 89 | ||
| GenBank mRNA | BC071361.1 | - | 6 - 7 | 959 | 89 | |
| KM655576.1 | - | 5 - 6 | 854 | 89 | ||
More Information
Set Membership: |
|

calua
Genomic Map