Assay Details
| Gene Symbol: uox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002332.1 | NP_001002332.1 | 6 - 7 | 763 | 133 | |
| GenBank mRNA | BC076466.1 | - | 6 - 7 | 763 | 133 | |
| BC164061.1 | - | 6 - 7 | 772 | 133 | ||
More Information
Set Membership: |
|

uox
Genomic Map