Assay Details
| Gene Symbol: pdha1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002399.1 | NP_001002399.1 | 3 - 4 | 364 | 83 | |
| XM_005162686.4 | XP_005162743.1 | 4 - 5 | 365 | 83 | ||
| GenBank mRNA | BC076185.1 | - | 3 - 4 | 364 | 83 | |
More Information
Set Membership: |
|

pdha1b
Genomic Map