Assay Details
| Gene Symbol: dgat1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002458.1 | NP_001002458.1 | 14 - 15 | 1432 | 75 | |
| XM_021466677.1 | XP_021322352.1 | 13 - 14 | 1062 | 75 | ||
| GenBank mRNA | BC076012.1 | - | 14 - 15 | 1432 | 75 | |
More Information
Set Membership: |
|

dgat1b
Genomic Map