Assay Details
| Gene Symbol: atp2b3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002472.3 | NP_001002472.2 | 9 - 10 | 2315 | 77 | |
| NM_001365433.1 | NP_001352362.1 | 10 - 11 | 2381 | 77 | ||
| XM_005166818.4 | XP_005166875.1 | 10 - 11 | 2300 | 77 | ||
| XM_005166819.4 | XP_005166876.1 | 9 - 10 | 2233 | 77 | ||
| XM_005166820.4 | XP_005166877.1 | 10 - 11 | 2300 | 77 | ||
| XM_005166822.4 | XP_005166879.1 | 10 - 11 | 2302 | 77 | ||
| XM_021478239.1 | XP_021333914.1 | 9 - 10 | 2233 | 77 | ||
| XM_021478240.1 | XP_021333915.1 | 9 - 10 | 2235 | 77 | ||
| GenBank mRNA | BC076342.1 | - | 8 - 9 | 1668 | 77 | |
| EU559283.1 | - | 8 - 9 | 1664 | 77 | ||
More Information
Set Membership: |
|

atp2b3a
Genomic Map