Assay Details
| Gene Symbol: itm2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002549.1 | NP_001002549.1 | 3 - 4 | 541 | 70 | |
| GenBank mRNA | BC076257.1 | - | 3 - 4 | 541 | 70 | |
More Information
Set Membership: |
|

itm2ca
Genomic Map