Assay Details
| Gene Symbol: pigh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002565.2 | NP_001002565.1 | 4 - 5 | 672 | 89 | |
| GenBank mRNA | BC076235.1 | - | 4 - 5 | 673 | 89 | |
More Information
Set Membership: |
|

pigh
Genomic Map