Assay Details
| Gene Symbol: hsd17b14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003521.2 | NP_001003521.1 | 3 - 4 | 347 | 97 | |
| GenBank mRNA | BC077136.1 | - | 3 - 4 | 351 | 97 | |
More Information
Set Membership: |
|

hsd17b14
Genomic Map