Assay Details
| Gene Symbol: immp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003755.2 | NP_001003755.1 | 5 - 6 | 446 | 63 | |
| GenBank mRNA | BC078193.1 | - | 5 - 6 | 450 | 63 | |
More Information
Set Membership: |
|

immp2l
Genomic Map