Assay Details
| Gene Symbol: cratb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004594.2 | NP_001004594.2 | 12 - 13 | 1686 | 70 | |
| GenBank mRNA | BC081604.1 | - | 12 - 13 | 1613 | 70 | |
More Information
Set Membership: |
|

cratb
Genomic Map