Assay Details
| Gene Symbol: cratb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004594.2 | NP_001004594.2 | 14 - 15 | 1888 | 66 | |
| GenBank mRNA | BC081604.1 | - | 14 - 15 | 1815 | 66 | |
More Information
Set Membership: |
|

cratb
Genomic Map