Assay Details
| Gene Symbol: cratb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004594.2 | NP_001004594.2 | 4 - 5 | 691 | 68 | |
| GenBank mRNA | BC081604.1 | - | 4 - 5 | 618 | 68 | |
More Information
Set Membership: |
|

cratb
Genomic Map