Assay Details
| Gene Symbol: cratb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004594.2 | NP_001004594.2 | 6 - 7 | 1024 | 149 | |
| GenBank mRNA | BC081604.1 | - | 6 - 7 | 951 | 149 | |
More Information
Set Membership: |
|

cratb
Genomic Map