Assay Details
| Gene Symbol: stk25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320056.1 | NP_001306985.1 | 3 - 4 | 583 | 87 | |
| XM_005165925.4 | XP_005165982.1 | 3 - 4 | 640 | 87 | ||
| GenBank mRNA | BC083536.1 | - | 3 - 4 | 549 | 87 | |
| EH435265.1 | - | 3 - 4 | 609 | 87 | ||
More Information
Set Membership: |
|

stk25a
Genomic Map