Assay Details
| Gene Symbol: bcl7bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006018.1 | NP_001006018.1 | 3 - 4 | 323 | 99 | |
| GenBank mRNA | BC083405.1 | - | 3 - 4 | 323 | 99 | |
| BC165913.1 | - | 3 - 4 | 303 | 99 | ||
More Information
Set Membership: |
|

bcl7bb
Genomic Map