Assay Details
| Gene Symbol: pcyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006037.1 | NP_001006037.1 | 1 - 2 | 223 | 65 | |
| GenBank mRNA | BC083378.1 | - | 1 - 2 | 223 | 65 | |
| BC164006.1 | - | 1 - 2 | 145 | 65 | ||
More Information
Set Membership: |
|

pcyt2
Genomic Map