Assay Details
| Gene Symbol: rab34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006094.1 | NP_001006094.1 | 4 - 5 | 371 | 124 | |
| GenBank mRNA | BC083224.1 | - | 4 - 5 | 371 | 124 | |
| BC163990.1 | - | 3 - 4 | 238 | 124 | ||
More Information
Set Membership: |
|

rab34a
Genomic Map