Assay Details
| Gene Symbol: rab34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006094.1 | NP_001006094.1 | 6 - 7 | 553 | 85 | |
| XM_005157422.4 | XP_005157479.1 | 6 - 7 | 605 | 85 | ||
| GenBank mRNA | BC083224.1 | - | 6 - 7 | 553 | 85 | |
| BC163990.1 | - | 5 - 6 | 420 | 85 | ||
More Information
Set Membership: |
|

rab34a
Genomic Map