Assay Details
| Gene Symbol: rab34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006094.1 | NP_001006094.1 | 9 - 10 | 773 | 82 | |
| XM_005157422.4 | XP_005157479.1 | 9 - 10 | 825 | 82 | ||
| GenBank mRNA | BC083224.1 | - | 9 - 10 | 773 | 82 | |
| BC163990.1 | - | 8 - 9 | 640 | 82 | ||
More Information
Set Membership: |
|

rab34a
Genomic Map