Assay Details
| Gene Symbol: dpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007152.1 | NP_001007153.1 | 2 - 3 | 211 | 68 | |
| XM_005165302.4 | XP_005165359.1 | 2 - 3 | 254 | 68 | ||
| XM_009301640.3 | XP_009299915.1 | 2 - 3 | 403 | 68 | ||
| XM_021476126.1 | XP_021331801.1 | 2 - 3 | 184 | 68 | ||
| GenBank mRNA | BC083281.1 | - | 2 - 3 | 211 | 68 | |
| BC165788.1 | - | 2 - 3 | 233 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021476126.1 transcripts.
Set Membership: |
|

dpf2
Genomic Map