Assay Details
| Gene Symbol: amh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007779.1 | NP_001007780.1 | 6 - 7 | 1228 | 72 | |
| XM_009296234.3 | XP_009294509.1 | 6 - 7 | 1228 | 72 | ||
| GenBank mRNA | AY677080.2 | - | 6 - 7 | 1311 | 72 | |
| AY721604.1 | - | 6 - 7 | 1228 | 72 | ||
| AY881649.1 | - | 6 - 7 | 1228 | 72 | ||
More Information
Set Membership: |
|

amh
Genomic Map