Assay Details
| Gene Symbol: gpm6bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013334.2 | NP_001013352.2 | 2 - 3 | 257 | 92 | |
| NM_001362553.1 | NP_001349482.1 | 2 - 3 | 412 | 92 | ||
| NM_001362554.1 | NP_001349483.1 | 2 - 3 | 257 | 92 | ||
| GenBank mRNA | BC090796.1 | - | 1 - 2 | 197 | 92 | |
| BC164613.1 | - | 1 - 2 | 218 | 92 | ||
More Information
Set Membership: |
|

gpm6bb
Genomic Map