Assay Details
| Gene Symbol: plcxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014300.1 | NP_001014322.1 | 1 - 2 | 149 | 79 | |
| GenBank mRNA | BC091866.1 | - | 1 - 2 | 149 | 79 | |
| BC154361.1 | - | 1 - 2 | 158 | 79 | ||
More Information
Set Membership: |
|

plcxd3
Genomic Map