Assay Details
| Gene Symbol: sema6dl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017682.2 | NP_001017682.2 | 10 - 11 | 1078 | 73 | |
| XM_005173648.4 | XP_005173705.1 | 10 - 11 | 1196 | 73 | ||
| XM_005173649.4 | XP_005173706.1 | 10 - 11 | 1196 | 73 | ||
| XM_005173650.4 | XP_005173707.1 | 10 - 11 | 1196 | 73 | ||
| XM_009293805.3 | XP_009292080.1 | 10 - 11 | 1587 | 73 | ||
| XM_009293807.3 | XP_009292082.1 | 10 - 11 | 1196 | 73 | ||
| GenBank mRNA | AB212073.1 | - | 10 - 11 | 1078 | 73 | |
| AB212074.1 | - | 10 - 11 | 1078 | 73 | ||
| CU458858.1 | - | 9 - 10 | 984 | 73 | ||
| CU458860.1 | - | 9 - 10 | 978 | 73 | ||
More Information
Set Membership: |
|

sema6dl
Genomic Map