Assay Details
| Gene Symbol: sema6dl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017682.2 | NP_001017682.2 | 11 - 12 | 1199 | 84 | |
| XM_005173648.4 | XP_005173705.1 | 11 - 12 | 1317 | 84 | ||
| XM_005173649.4 | XP_005173706.1 | 11 - 12 | 1317 | 84 | ||
| XM_005173650.4 | XP_005173707.1 | 11 - 12 | 1317 | 84 | ||
| XM_009293805.3 | XP_009292080.1 | 11 - 12 | 1708 | 84 | ||
| XM_009293807.3 | XP_009292082.1 | 11 - 12 | 1317 | 84 | ||
| GenBank mRNA | AB212073.1 | - | 11 - 12 | 1199 | 84 | |
| AB212074.1 | - | 11 - 12 | 1199 | 84 | ||
More Information
Set Membership: |
|

sema6dl
Genomic Map