Assay Details
| Gene Symbol: sema6dl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017682.2 | NP_001017682.2 | 16 - 17 | 1806 | 95 | |
| XM_005173648.4 | XP_005173705.1 | 16 - 17 | 1924 | 95 | ||
| XM_009293805.3 | XP_009292080.1 | 16 - 17 | 2315 | 95 | ||
| XM_009293807.3 | XP_009292082.1 | 16 - 17 | 1924 | 95 | ||
| GenBank mRNA | AB212073.1 | - | 16 - 17 | 1806 | 95 | |
More Information
Set Membership: |
|

sema6dl
Genomic Map