Assay Details
| Gene Symbol: sema6dl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017682.2 | NP_001017682.2 | 17 - 18 | 1871 | 83 | |
| GenBank mRNA | AB212073.1 | - | 17 - 18 | 1871 | 83 | |
More Information
Set Membership: |
|

sema6dl
Genomic Map