Assay Details
| Gene Symbol: sema6dl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017682.2 | NP_001017682.2 | 3 - 4 | 324 | 68 | |
| XM_005173648.4 | XP_005173705.1 | 3 - 4 | 442 | 68 | ||
| XM_005173649.4 | XP_005173706.1 | 3 - 4 | 442 | 68 | ||
| XM_005173650.4 | XP_005173707.1 | 3 - 4 | 442 | 68 | ||
| XM_009293805.3 | XP_009292080.1 | 3 - 4 | 833 | 68 | ||
| XM_009293807.3 | XP_009292082.1 | 3 - 4 | 442 | 68 | ||
| GenBank mRNA | AB212073.1 | - | 3 - 4 | 324 | 68 | |
| AB212074.1 | - | 3 - 4 | 324 | 68 | ||
| CU458858.1 | - | 2 - 3 | 230 | 68 | ||
| CU458860.1 | - | 2 - 3 | 224 | 68 | ||
| FJ209036.1 | - | 2 - 3 | 265 | 68 | ||
More Information
Set Membership: |
|

sema6dl
Genomic Map