Assay Details
| Gene Symbol: f3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017728.2 | NP_001017728.2 | 1 - 2 | 180 | 71 | |
| GenBank mRNA | BC093227.1 | - | 1 - 2 | 173 | 71 | |
| EH551524.1 | - | 1 - 2 | 219 | 71 | ||
More Information
Set Membership: |
|

f3b
Genomic Map