Assay Details
| Gene Symbol: f3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017728.2 | NP_001017728.2 | 2 - 3 | 302 | 76 | |
| GenBank mRNA | BC093227.1 | - | 2 - 3 | 295 | 76 | |
| EH551524.1 | - | 2 - 3 | 341 | 76 | ||
More Information
Set Membership: |
|

f3b
Genomic Map