Assay Details
| Gene Symbol: dnmt3ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018140.2 | NP_001018150.1 | 10 - 11 | 1626 | 60 | |
| NM_001328167.1 | NP_001315096.1 | 15 - 16 | 2175 | 60 | ||
| XM_005158845.4 | XP_005158902.1 | 10 - 11 | 1540 | 60 | ||
| XM_009293199.3 | XP_009291474.1 | 14 - 15 | 2062 | 60 | ||
| XM_009293200.3 | XP_009291475.1 | 10 - 11 | 1633 | 60 | ||
| XM_021467161.1 | XP_021322836.1 | 15 - 16 | 2315 | 60 | ||
| XM_021467162.1 | XP_021322837.1 | 15 - 16 | 2225 | 60 | ||
| XM_021467163.1 | XP_021322838.1 | 15 - 16 | 2226 | 60 | ||
| GenBank mRNA | AB196917.1 | - | 10 - 11 | 1628 | 60 | |
| BC162582.1 | - | 10 - 11 | 1538 | 60 | ||
More Information
Set Membership: |
|

dnmt3ab
Genomic Map