Assay Details
| Gene Symbol: myh11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024448.1 | NP_001019619.1 | 29 - 30 | 4066 | 93 | |
| XM_021476875.1 | XP_021332550.1 | 30 - 31 | 4119 | 93 | ||
| XM_021476876.1 | XP_021332551.1 | 30 - 31 | 4119 | 93 | ||
| GenBank mRNA | BC162152.1 | - | 29 - 30 | 4030 | 93 | |
| DQ022376.1 | - | 29 - 30 | 4066 | 93 | ||
More Information
Set Membership: |
|

myh11a
Genomic Map