Assay Details
| Gene Symbol: myh11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024448.1 | NP_001019619.1 | 6 - 7 | 838 | 86 | |
| XM_021476875.1 | XP_021332550.1 | 7 - 8 | 891 | 86 | ||
| XM_021476876.1 | XP_021332551.1 | 7 - 8 | 891 | 86 | ||
| GenBank mRNA | BC162152.1 | - | 6 - 7 | 802 | 86 | |
| DQ022376.1 | - | 6 - 7 | 838 | 86 | ||
More Information
Set Membership: |
|

myh11a
Genomic Map