Assay Details
| Gene Symbol: prickle1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030098.2 | NP_001025269.1 | 6 - 7 | 967 | 96 | |
| XM_005164634.4 | XP_005164691.1 | 6 - 7 | 1233 | 96 | ||
More Information
Set Membership: |
|

prickle1b
Genomic Map