Assay Details
| Gene Symbol: otofa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030112.2 | NP_001025283.1 | 14 - 15 | 1588 | 83 | |
| XM_009294732.3 | XP_009293007.1 | 14 - 15 | 1887 | 83 | ||
| XM_009294733.3 | XP_009293008.1 | 14 - 15 | 1888 | 83 | ||
| XM_009294734.3 | XP_009293009.1 | 14 - 15 | 1887 | 83 | ||
| XM_009294736.3 | XP_009293011.1 | 14 - 15 | 1887 | 83 | ||
| XM_009294738.3 | XP_009293013.1 | 14 - 15 | 1888 | 83 | ||
| XM_009294739.3 | XP_009293014.1 | 14 - 15 | 1887 | 83 | ||
| XM_009294740.3 | XP_009293015.1 | 14 - 15 | 1887 | 83 | ||
| XM_009294742.3 | XP_009293017.1 | 14 - 15 | 1887 | 83 | ||
| XM_009294744.3 | XP_009293019.1 | 14 - 15 | 1887 | 83 | ||
| XM_021468441.1 | XP_021324116.1 | 14 - 15 | 1887 | 83 | ||
| XM_021468442.1 | XP_021324117.1 | 14 - 15 | 1887 | 83 | ||
| XM_021468443.1 | XP_021324118.1 | 14 - 15 | 1887 | 83 | ||
More Information
Set Membership: |
|

otofa
Genomic Map