Assay Details
| Gene Symbol: kdm6bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030178.2 | NP_001025349.2 | 10 - 11 | 4027 | 129 | |
| XM_005172621.4 | XP_005172678.1 | 10 - 11 | 4082 | 129 | ||
| XM_005172623.4 | XP_005172680.1 | 10 - 11 | 4078 | 129 | ||
| XM_021479186.1 | XP_021334861.1 | 10 - 11 | 3990 | 129 | ||
More Information
Set Membership: |
|

kdm6bb
Genomic Map