Assay Details
| Gene Symbol: per1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030183.1 | NP_001025354.1 | 10 - 11 | 1666 | 63 | |
| XM_005172625.4 | XP_005172682.1 | 11 - 12 | 2077 | 63 | ||
| XM_005172627.4 | XP_005172684.1 | 11 - 12 | 2077 | 63 | ||
| XM_021479187.1 | XP_021334862.1 | 11 - 12 | 2077 | 63 | ||
| GenBank mRNA | BC163543.1 | - | 10 - 11 | 1735 | 63 | |
More Information
Set Membership: |
|

per1a
Genomic Map