Assay Details
| Gene Symbol: per1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030183.1 | NP_001025354.1 | 6 - 7 | 1054 | 73 | |
| XM_005172625.4 | XP_005172682.1 | 7 - 8 | 1465 | 73 | ||
| XM_005172626.4 | XP_005172683.1 | 7 - 8 | 1465 | 73 | ||
| XM_005172627.4 | XP_005172684.1 | 7 - 8 | 1465 | 73 | ||
| XM_021479187.1 | XP_021334862.1 | 7 - 8 | 1465 | 73 | ||
| GenBank mRNA | BC163543.1 | - | 6 - 7 | 1123 | 73 | |
More Information
Set Membership: |
|

per1a
Genomic Map