Assay Details
| Gene Symbol: per1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030183.1 | NP_001025354.1 | 8 - 9 | 1312 | 82 | |
| XM_005172625.4 | XP_005172682.1 | 9 - 10 | 1723 | 82 | ||
| XM_005172626.4 | XP_005172683.1 | 9 - 10 | 1723 | 82 | ||
| XM_005172627.4 | XP_005172684.1 | 9 - 10 | 1723 | 82 | ||
| XM_021479187.1 | XP_021334862.1 | 9 - 10 | 1723 | 82 | ||
| GenBank mRNA | BC163543.1 | - | 8 - 9 | 1381 | 82 | |
More Information
Set Membership: |
|

per1a
Genomic Map