Assay Details
| Gene Symbol: tnfaip2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030216.1 | NP_001025387.1 | 7 - 8 | 1398 | 147 | |
| XM_005160467.4 | XP_005160524.1 | 8 - 9 | 1478 | 147 | ||
| GenBank mRNA | BC114226.1 | - | 7 - 8 | 1401 | 147 | |
More Information
Set Membership: |
|

tnfaip2a
Genomic Map