Assay Details
| Gene Symbol: tnfaip2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030216.1 | NP_001025387.1 | 9 - 10 | 1616 | 91 | |
| XM_005160467.4 | XP_005160524.1 | 10 - 11 | 1696 | 91 | ||
| GenBank mRNA | BC114226.1 | - | 9 - 10 | 1619 | 91 | |
More Information
Set Membership: |
|

tnfaip2a
Genomic Map